Learn More
Medchemexpress LLC L-Glutamine-15N2 | 204451-48-9 | 148.13 | 5 MG
SDP
Catalog No. 5000390209
Encompass_Preferred
L-Glutamine-15N2 is the 15N-labeled L-Glutamine, a non-essential amino acid abundant throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells. Stable heavy isotopes of elements like carbon and nitrogen are incorporated into molecules, serving as tracers for quantitative analysis during drug development.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-90209 Supplier Medchemexpress LLC Supplier No. HYN0390S85MG
L-Glutamine-15N2 is the 15N-labeled L-Glutamine, a non-essential amino acid abundant throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells. Stable heavy isotopes of elements like carbon and nitrogen are incorporated into molecules, serving as tracers for quantitative analysis during drug development.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS