Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech L-Glutamine 200 mM (100 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0480100
Encompass_Preferred
L-Glutamine 200 mM. QUANTITY: 100 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: Negative 20 degrees Celsius. DESCRIPTION: L-Glutamine is an essential amino acid required by all mammalian and insect cells grown in culture. It serves as an additional source of carbon and energy and in particular is used for nucleic acid synthesis. Optimal cell performance usually requires supplementation of the medium with L-Glutamine prior to use.
Catalog No. 50-238-8525