Learn More
ATCC L-Glutamine Solution, 200 mM, 100mL, Frozen
Supplier:  ATCC 302214/DEL
Encompass
L-Glutamine Solution is added as a supplement to cell culture media. L-Glutamine is an essential amino acid required by virtually all mammalian and insect cells grown in culture. L-Glutamine is also more labile in cell culture solutions than other amino acids. The rate and extent of L-glutamine degradation are related to storage temperatures, age of the product, and pH. For use by FOR-PROFIT companies only; NON-PROFIT companies use 50188274NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-188-274FP