Learn More
Quality Biological Inc L-Glutamine 200 mM, PK 4 X 100 mL
SDP
Supplier:  Quality Biological Inc 118084721
Encompass_Preferred
L-glutamine is a cell culture supplement, and an essential amnio acid. ¿ L-glutamine is required in cell culture for cellular growth. ¿ 100X concentration ¿ Applications: used in cell culture as an energy source in addition to carbohydrates (i.e. glucose, sucrose, maltose, etc.). ¿ L-glutamine participates in the formation of purine and pyrimidine nucleotides, amino sugars, gluthione, L-glutamate and other amino acids, as well as in protein synthesis and glucose production. ¿ Storage Conditions: -10¿C to -20¿C ¿ Shipping Condition: on dry ice ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 240 months
Catalog No. 50-983-211