Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamine | 56-85-9 | MFCD00008044 | 146.14 g/mol | C5H10N2O3 | 10 MG
SDP

Catalog No. 5000295735
Encompass_Preferred

L-Glutamine (Standard) is an analytical reference standard of L-glutamine (L-glutamic acid 5-amide) supplied as a solid for research and analytical applications. It is intended for qualitative and quantitative method development and validation and is commonly used in HPLC, GC, and MS workflows. Documentation such as SDS and COA/HNMR reports are available on request.

  • Analytical reference standard for method development and validation.
  • Suitable for qualitative and quantitative HPLC, GC, and MS analyses.
  • CAS number 56-85-9 for unambiguous identification.
  • Molecular formula C5H10N2O3; molecular weight 146.14 g/mol.
  • Provided in small reference pack sizes (for example, 10 MG) for laboratory use.
  • SDS and handling instructions available; COA or HNMR report available on request.

Catalog No. 50-002-95735 Supplier Medchemexpress LLC Supplier No. HYN0390R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamine (Standard) is an analytical reference standard of L-glutamine (L-glutamic acid 5-amide) supplied as a solid for research and analytical applications. It is intended for qualitative and quantitative method development and validation and is commonly used in HPLC, GC, and MS workflows. Documentation such as SDS and COA/HNMR reports are available on request.

  • Analytical reference standard for method development and validation.
  • Suitable for qualitative and quantitative HPLC, GC, and MS analyses.
  • CAS number 56-85-9 for unambiguous identification.
  • Molecular formula C5H10N2O3; molecular weight 146.14 g/mol.
  • Provided in small reference pack sizes (for example, 10 MG) for laboratory use.
  • SDS and handling instructions available; COA or HNMR report available on request.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.