Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamine, 99-100%, MP Biomedicals

Catalog No. ICN19467890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19467890 500 g
ICN19467825 25 g
ICN19467880 100 g
ICN19467883 250 g
ICN19467891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19467890 Supplier MP Biomedicals, Inc Supplier No. 0219467890
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Quantity 500 g
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Glutamine, 99-100%
Recommended Storage Room temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.