Learn More
Medchemexpress LLC L-Glutamine-d5 | 14341-78-7 | 151.18 | 5 MG
SDP
Catalog No. 5000389925
Encompass_Preferred
L-Glutamine-d5 is the deuterium labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid present abundantly throughout the body and involved in many metabolic processes. L-Glutamine provides a source of carbons for oxidation in some cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • For research use only.
Catalog No. 50-003-89925 Supplier Medchemexpress LLC Supplier No. HYN0390S25MG
L-Glutamine-d5 is the deuterium labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid present abundantly throughout the body and involved in many metabolic processes. L-Glutamine provides a source of carbons for oxidation in some cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • For research use only.