Learn More
Medchemexpress LLC L-Glutamine-d5 (L-Glutamic acid 5-amide-d5) | 14341-78-7 | 151.18 | 1 MG
SDP
Catalog No. 5000389978
Encompass_Preferred
L-Glutamine-d5 is a deuterium-labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid found abundantly throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89978 Supplier Medchemexpress LLC Supplier No. HYN0390S21MG
L-Glutamine-d5 is a deuterium-labeled L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid found abundantly throughout the body and involved in many metabolic processes. It provides a source of carbons for oxidation in some cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.