Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glutamine-d5 (L-glutamic acid 5-amide-d5) | 14341-78-7 | MFCD00144600 | 99.8% | 151.18 g/mol | C5H5D5N2O3 | 10 MG
SDP

Catalog No. 5000295644
Encompass_Preferred

L-Glutamine-d5 is a deuterium-labeled analogue of L-glutamine supplied as a stable isotope standard for research applications, including metabolic studies and mass spectrometry. It is provided as a solid with high chemical and isotopic purity and is intended for research use only (not for human use).

  • Deuterium-labeled isotopologue of L-glutamine.
  • High purity (99.8%).
  • Molecular weight approximately 151.18 g/mol.
  • Chemical formula: C5H5D5N2O3.
  • Supplied in small milligram pack sizes suitable for analytical standards.
  • Store sealed, away from moisture and light; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).

Catalog No. 50-002-95644 Supplier Medchemexpress LLC Supplier No. HYN0390S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

L-Glutamine-d5 is a deuterium-labeled analogue of L-glutamine supplied as a stable isotope standard for research applications, including metabolic studies and mass spectrometry. It is provided as a solid with high chemical and isotopic purity and is intended for research use only (not for human use).

  • Deuterium-labeled isotopologue of L-glutamine.
  • High purity (99.8%).
  • Molecular weight approximately 151.18 g/mol.
  • Chemical formula: C5H5D5N2O3.
  • Supplied in small milligram pack sizes suitable for analytical standards.
  • Store sealed, away from moisture and light; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.