Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamine, MP Biomedicals

Catalog No. MP210180691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210180691 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210180691 Supplier MP Biomedicals, Inc Supplier No. 0210180691
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity powder for use in research applications.

  • L-glutamine is an essential amino acid, which serves as a major energy source for cells in culture
  • L-glutamine is added for optimal cell performance in a culture medium
  • The utilization of glutamine under carbon starvation conditions by cultured Saccharomyces cerevisiae and its relation to the cytoplasmic compartmentation of the transcriptional activator Gln3 has been studied
  • The use of glutamine in cultured rat islets to probe a-ketoisocaproate oxidation has been investigated

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185°C (literature)
Color White
Linear Formula C5H10N2O3
Solubility Information Soluble in water.
Quantity 1 kg
Formula Weight 146.1
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material L-GLUTAMINE
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.