Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Glutamine-N,N2-15N2 | 204451-48-9 | 148.13 | 25 MG
SDP

Catalog No. 5000389963
Encompass_Preferred

L-Glutamine-15N2 is the 15N-labeled version of L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid that is abundant throughout the body and involved in many metabolic processes, providing a source of carbons for oxidation in some cells. This product is for research use only and not sold to patients.

  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes of nitrogen (15N).
  • Purity: 99.10%.
  • Documentation: data sheet, certificate of analysis, safety data sheet, and handling instructions are available.
  • Available in various in-stock sizes.

Catalog No. 50-003-89963 Supplier Medchemexpress LLC Supplier No. 25MGHYN0390S8
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Glutamine-15N2 is the 15N-labeled version of L-Glutamine. L-Glutamine (L-Glutamic acid 5-amide) is a non-essential amino acid that is abundant throughout the body and involved in many metabolic processes, providing a source of carbons for oxidation in some cells. This product is for research use only and not sold to patients.

  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes of nitrogen (15N).
  • Purity: 99.10%.
  • Documentation: data sheet, certificate of analysis, safety data sheet, and handling instructions are available.
  • Available in various in-stock sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.