Learn More
Medchemexpress LLC L-Glutamine (Standard) | 56-85-9 | 146.14 | 100 MG
SDP
Catalog No. 5000388981
Encompass_Preferred
L-Glutamine (Standard) is an analytical standard for L-Glutamine, a non-essential amino acid involved in many metabolic processes. It provides a source of carbons for oxidation in various cells and is intended for research and analytical applications.
  • Used as a reference standard assay.
  • Commonly used in qualitative research experiments.
  • Commonly used in quantitative research experiments.
  • Commonly used in methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-003-88981 Supplier Medchemexpress LLC Supplier No. HYN0390R100MG
L-Glutamine (Standard) is an analytical standard for L-Glutamine, a non-essential amino acid involved in many metabolic processes. It provides a source of carbons for oxidation in various cells and is intended for research and analytical applications.
  • Used as a reference standard assay.
  • Commonly used in qualitative research experiments.
  • Commonly used in quantitative research experiments.
  • Commonly used in methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.