Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamine, USP Grade, MP Biomedicals™

Catalog No. 0219960891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-199-60891 1000 g
02-199-60880 100 g
02-199-60890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-199-60891 Supplier MP Biomedicals, Inc Supplier No. 0219960891
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym (S)-2,5-Diamino-5-oxopentanoic acid; L-Glutamic acid 5-amide
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185.5°C
pH 5 to 6 (at 14.6 g/L, 25°C)
Color White
Beilstein 1723797
Density 1.364 g/cm3
Quantity 1000 g
Optical Rotation (40 mg/mL aq. soln. (at 20°C) +6.3° to +7.3°)
Grade USP
Physical Form Powder
Chemical Name or Material L-Glutamine, USP

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.