Learn More
Sigma Aldrich Fine Chemicals Biosciences L Glutamne 13C515N2 98 at100MG
Catalog No. 502903064
Encompass
Glutamine is an amino acid with an uncharged polar side chain. It can be catabolized to glutamate and ammonia. Glutamine is the most abundant amino acid in the central nervous system. Isotope labeled amino acids are required for the production of isotopically labeled proteins.
Catalog No. 50-290-3064 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 607983100MG
Glutamine is an amino acid with an uncharged polar side chain. It can be catabolized to glutamate and ammonia. Glutamine is the most abundant amino acid in the central nervous system. Isotope labeled amino acids are required for the production of isotopically labeled proteins.