Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-γ-glutamyl-L-arginine | 31106-03-3 | 303.31 | 50 MG
SDP

Catalog No. 5000417047
Encompass_Preferred

gamma-Glutamylarginine (L-γ-Glutamyl-L-arginine) is an γ-glutamic acid oligopeptide that can be isolated from *Panax ginseng*. This product is for research use only.

  • Isolated from *Panax ginseng*
  • An γ-glutamic acid oligopeptide
  • Appearance: solid
  • Color: white to off-white
  • Purity: 97.70%

Catalog No. 50-004-17047 Supplier Medchemexpress LLC Supplier No. HYN1047750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

gamma-Glutamylarginine (L-γ-Glutamyl-L-arginine) is an γ-glutamic acid oligopeptide that can be isolated from *Panax ginseng*. This product is for research use only.

  • Isolated from *Panax ginseng*
  • An γ-glutamic acid oligopeptide
  • Appearance: solid
  • Color: white to off-white
  • Purity: 97.70%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.