Learn More
Medchemexpress LLC HY-D0187 1g Medchemexpress, L-Glutathione reduced CAS:70-18-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD01871G
Encompass_Preferred
Medchemexpress, HY-D0187 1g L-Glutathione reduced CAS:70-18-8 L-Glutathione reduced (GSH; γ-L-Glutamyl-L-cysteinyl-glycine) is an endogenous antioxidant and is capable of scavenging oxygen-derived free radicals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3155