Learn More
Apexbio Technology LLC L-Glutathione Reduced(Synonyms: Reduced Glutathione, GSH, Gamma-L-Glutamyl-L-cysteinylglycine, L-Glutathione), 10mM (in 1mL H2O), CAS: 70-18-8.
SDP
Catalog No. 5000060207
Encompass_Preferred
L-Glutathione Reduced (CAS 70-18-8) is an endogenous thiol-containing tripeptide antioxidant found ubiquitously in living cells It exerts biological effects primarily by scavenging reactive oxygen species (ROS) and maintaining the redox balance within cells By neutralizing oxygen radicals and regenerating other antioxidants including vitamin E and ascorbic acid it mitigates oxidative stress implicated in conditions such as cancer cardiovascular diseases and inflammation In biomedical research L-Glutathione Reduced also serves as a substrate for glutathione S-transferase (GST) affinity purification methods and as a biomarker in oxidative stress studies
Catalog No. 50-000-60207 Supplier Apexbio Technology LLC Supplier No. B77755.1
L-Glutathione Reduced (CAS 70-18-8) is an endogenous thiol-containing tripeptide antioxidant found ubiquitously in living cells It exerts biological effects primarily by scavenging reactive oxygen species (ROS) and maintaining the redox balance within cells By neutralizing oxygen radicals and regenerating other antioxidants including vitamin E and ascorbic acid it mitigates oxidative stress implicated in conditions such as cancer cardiovascular diseases and inflammation In biomedical research L-Glutathione Reduced also serves as a substrate for glutathione S-transferase (GST) affinity purification methods and as a biomarker in oxidative stress studies