Learn More
Gold Biotechnology Inc L Glutathione Reduced 50 g
SDP
Catalog No. 5000562036
Encompass_Preferred
L-Glutathione is a co-substrate to glutathione transferase found in plants and animals and is involved in the elimination of free radicals resulting from both cellular respiratory and metabolic activities
Catalog No. 50-005-62036 Supplier Gold Biotechnology Inc Supplier No. G15550
L-Glutathione is a co-substrate to glutathione transferase found in plants and animals and is involved in the elimination of free radicals resulting from both cellular respiratory and metabolic activities