Learn More
AdipoGen L-Glutathione reduced
SDP
Supplier:  AdipoGen CDXG0005G005
Encompass_Preferred
Chemical. CAS 70-18-8. Formula C10H17N3O6S. MW 307.32. Synthetic. Major tripeptide widely distributed in both plants and animals. Endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst. Regulates activity of the redox sensitive transcription factor NF-kappaB. Cytoprotective. Serves as a nucleophilic co-substrate to glutathione transferases in the detoxification of xenobiotics and is an essential electron donor to glutathione peroxidases in the reduction of hydroperoxides. Involved in amino acid transport and maintenance of protein sulfhydryl reduction status. Posseses several metabolic, regulatory and protective functions. Forms disulfide bonds with cysteine residues in proteins.
Catalog No. 50-256-0848