Learn More
Enzo Life Sciences L-Glutathione (reduced form) (5g). CAS: 70-18-8
SDP
Supplier:  Enzo Life Sciences ALX157002G005
Encompass_Preferred
Alternative name: GSH, gamma-Glu-Cys-Gly-OH. Formula: C10H17N3O6S. MW: 307.3. Purity: ≥99%. Appearance: White to off-white solid. Formulation: Free acid. MI: 14: 4475. Solubility: Soluble in water. Long Term Storage: +4°C. Handling: Protect from light and moisture.
Catalog No. 50-200-7817