Learn More
Alkali Scientific L Glutathione, reduced, free acid, 25 G
SDP
Catalog No. 5000660116
Encompass_Preferred
L-Glutathione, Reduced, Free Acid, 25g, is an essential antioxidant used in biochemical and molecular biology research. It plays a critical role in cellular defense against oxidative stress by neutralizing free radicals and reactive oxygen species (ROS). L-Glutathione is also involved in various cellular processes, including DNA synthesis, repair, and protein synthesis. It is commonly used in enzyme assays, cell culture experiments, and as a reagent in redox studies. The reduced form is the biologically active version of glutathione, and this product is ideal for research involving oxidative stress, detoxification, and other cellular mechanisms.
Catalog No. 50-006-60116 Supplier Alkali Scientific Supplier No. LUT001CP
L-Glutathione, Reduced, Free Acid, 25g, is an essential antioxidant used in biochemical and molecular biology research. It plays a critical role in cellular defense against oxidative stress by neutralizing free radicals and reactive oxygen species (ROS). L-Glutathione is also involved in various cellular processes, including DNA synthesis, repair, and protein synthesis. It is commonly used in enzyme assays, cell culture experiments, and as a reagent in redox studies. The reduced form is the biologically active version of glutathione, and this product is ideal for research involving oxidative stress, detoxification, and other cellular mechanisms.