Learn More
Medchemexpress LLC L-Glutathione reduced (Standard) | 70-18-8 | 99.7% | 250 MG
SDP
Catalog No. 5000414321
Encompass_Preferred
L-Glutathione reduced (Standard) is the analytical standard of L-Glutathione reduced. This product is intended for research and analytical applications. L-Glutathione reduced (GSH; γ-L-Glutamyl-L-cysteinyl-glycine) is an endogenous antioxidant and is capable of scavenging oxygen-derived free radicals. The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Intended for research and analytical applications.
  • Endogenous antioxidant.
  • Capable of scavenging oxygen-derived free radicals.
  • Grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-14321 Supplier Medchemexpress LLC Supplier No. 250MGHYD0187R
L-Glutathione reduced (Standard) is the analytical standard of L-Glutathione reduced. This product is intended for research and analytical applications. L-Glutathione reduced (GSH; γ-L-Glutamyl-L-cysteinyl-glycine) is an endogenous antioxidant and is capable of scavenging oxygen-derived free radicals. The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Intended for research and analytical applications.
  • Endogenous antioxidant.
  • Capable of scavenging oxygen-derived free radicals.
  • Grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.