Learn More
Medchemexpress LLC L-Glyceric acid sodium | 146298-95-5 | 99.0% | 128.06 | 10 MG
SDP
Catalog No. 5000436957
Encompass_Preferred
L-Glyceric acid sodium is a urinary metabolite that accumulates in the rare inherited metabolic disease L-glyceric aciduria. It is used to diagnose primary hyperoxaluria type 2 (PH2) and to distinguish PH1 from PH2 based on its excretion.
  • Urinary metabolite
  • Used to diagnose primary hyperoxaluria type 2 (PH2)
  • Helps distinguish PH1 from PH2
  • For research use only
Catalog No. 50-004-36957 Supplier Medchemexpress LLC Supplier No. HY113377A10MG
L-Glyceric acid sodium is a urinary metabolite that accumulates in the rare inherited metabolic disease L-glyceric aciduria. It is used to diagnose primary hyperoxaluria type 2 (PH2) and to distinguish PH1 from PH2 based on its excretion.
  • Urinary metabolite
  • Used to diagnose primary hyperoxaluria type 2 (PH2)
  • Helps distinguish PH1 from PH2
  • For research use only