Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-glyceric acid sodium | 146298-95-5 | MFCD28363841 | 99.0% | 128.06 g/mol | C3H5NaO4 | 5 MG
SDP

Catalog No. 5000264328
Encompass_Preferred

L-glyceric acid sodium is the sodium salt of L-glyceric acid, a urinary metabolite that accumulates in L-glyceric aciduria. Supplied as a high-purity analytical reference standard, it is intended for metabolic research and diagnostic applications requiring accurate reference material. The compound has formula C3H5NaO4, molecular weight 128.06 g/mol, and reported purity 99.0%.

  • Purity of 99.0%.
  • Molecular weight 128.06 g/mol.
  • Chemical formula C3H5NaO4.
  • Available in small milligram pack sizes for analytical use.
  • Intended as an analytical reference for metabolic and diagnostic studies involving L-glyceric aciduria.

Catalog No. 50-002-64328 Supplier Medchemexpress LLC Supplier No. HY113377A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-glyceric acid sodium is the sodium salt of L-glyceric acid, a urinary metabolite that accumulates in L-glyceric aciduria. Supplied as a high-purity analytical reference standard, it is intended for metabolic research and diagnostic applications requiring accurate reference material. The compound has formula C3H5NaO4, molecular weight 128.06 g/mol, and reported purity 99.0%.

  • Purity of 99.0%.
  • Molecular weight 128.06 g/mol.
  • Chemical formula C3H5NaO4.
  • Available in small milligram pack sizes for analytical use.
  • Intended as an analytical reference for metabolic and diagnostic studies involving L-glyceric aciduria.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.