Learn More
Medchemexpress LLC L-Glycero-D-mannoheptose | 4305-74-2 | MFCD00269999 | >98.0% | 210.18 | C7H14O7 | 1 MG
SDP
Catalog No. 5000281693
Encompass_Preferred
L-Glycero-D-mannoheptose is a research-grade biochemical reagent used in carbohydrate and bacterial lipopolysaccharide (LPS) studies. It is a seven-carbon sugar with formula C7H14O7, molecular weight 210.18, and CAS 4305-74-2. The compound is supplied in small, analytical pack sizes for use as a reagent or reference standard in assays and structural studies.
  • Seven-carbon sugar used in LPS and O-antigen research.
  • Suitable as a reagent or analytical reference standard.
  • Supplied in milligram pack sizes for small-scale assays.
  • Molecular formula C7H14O7 and molecular weight 210.18 aid in experimental calculations.
  • Provided for research use only; not for human consumption.
Catalog No. 50-002-81693 Supplier Medchemexpress LLC Supplier No. HYW4159161MG
L-Glycero-D-mannoheptose is a research-grade biochemical reagent used in carbohydrate and bacterial lipopolysaccharide (LPS) studies. It is a seven-carbon sugar with formula C7H14O7, molecular weight 210.18, and CAS 4305-74-2. The compound is supplied in small, analytical pack sizes for use as a reagent or reference standard in assays and structural studies.
  • Seven-carbon sugar used in LPS and O-antigen research.
  • Suitable as a reagent or analytical reference standard.
  • Supplied in milligram pack sizes for small-scale assays.
  • Molecular formula C7H14O7 and molecular weight 210.18 aid in experimental calculations.
  • Provided for research use only; not for human consumption.