Learn More
Medchemexpress LLC L-glycero-D-mannoheptose | 4305-74-2 | MFCD00269999 | >98.0% | 210.18 g/mol | C7H14O7 | 5 MG
SDP
Catalog No. 5000281040
Encompass_Preferred
L-Glycero-D-mannoheptose is a seven-carbon aldoheptose used as a biochemical assay reagent and research chemical. It is commonly applied in studies of bacterial lipopolysaccharide biosynthesis and carbohydrate research, providing a defined, high-purity material for analytical and preparative workflows.
  • Seven-carbon aldoheptose for biochemical assays and carbohydrate research.
  • High purity (>98%) suitable for analytical and preparative work.
  • Molecular weight 210.18 g/mol and formula C7H14O7 for accurate calculations.
  • Supplied in small pack sizes for research-scale experiments.
  • Useful in studies of lipopolysaccharide biosynthesis and bacterial carbohydrate pathways.
Catalog No. 50-002-81040 Supplier Medchemexpress LLC Supplier No. HYW4159165MG
L-Glycero-D-mannoheptose is a seven-carbon aldoheptose used as a biochemical assay reagent and research chemical. It is commonly applied in studies of bacterial lipopolysaccharide biosynthesis and carbohydrate research, providing a defined, high-purity material for analytical and preparative workflows.
  • Seven-carbon aldoheptose for biochemical assays and carbohydrate research.
  • High purity (>98%) suitable for analytical and preparative work.
  • Molecular weight 210.18 g/mol and formula C7H14O7 for accurate calculations.
  • Supplied in small pack sizes for research-scale experiments.
  • Useful in studies of lipopolysaccharide biosynthesis and bacterial carbohydrate pathways.