Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glycine 7-amido-4-methylcoumarin, Thermo Scientific Chemicals

Catalog No. AAJ65592MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65592MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65592MA Supplier Thermo Scientific Chemicals Supplier No. J65592MA
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12H12N2O3
MDL Number MFCD00077122
Synonym L-Gly-AMC; L-G-AMC

Specifications

Color White
Quantity 10 mg
Sensitivity Light sensitive
Solubility Information Soluble in DMSO or water
Formula Weight 232.23
Physical Form Solid
Chemical Name or Material L-Glycine 7-amido-4-methylcoumarin
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.