Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Guanosine | 26578-09-6 | 98.0% | 283.24 | 25 MG
SDP

Catalog No. 5000351869
Encompass_Preferred

L-Guanosine is the L-configuration of Guanosine. It is a purine nucleoside that exhibits anti-herpesvirus activity and can be used to prepare supramolecular hydrogel. This product ships at room temperature and should be stored solid at 4°C, protected from light. Solutions can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light.

  • Purine nucleoside
  • Exhibits anti-herpesvirus activity
  • Can be used to prepare supramolecular hydrogel
  • Ships at room temperature
  • Solid storage: 4°C, protect from light
  • In solvent storage: -80°C for 6 months or -20°C for 1 month, protect from light

Catalog No. 50-003-51869 Supplier Medchemexpress LLC Supplier No. 25MGHY138130
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Guanosine is the L-configuration of Guanosine. It is a purine nucleoside that exhibits anti-herpesvirus activity and can be used to prepare supramolecular hydrogel. This product ships at room temperature and should be stored solid at 4°C, protected from light. Solutions can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light.

  • Purine nucleoside
  • Exhibits anti-herpesvirus activity
  • Can be used to prepare supramolecular hydrogel
  • Ships at room temperature
  • Solid storage: 4°C, protect from light
  • In solvent storage: -80°C for 6 months or -20°C for 1 month, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.