Learn More
Medchemexpress LLC L-Gulono-1,4-lactone | 1128-23-0 | MFCD00064331 | 178.14 g/mol | C6H10O6 | 1 ML
SDP
Catalog No. 5000300801
Encompass_Preferred
L-Gulono-1,4-lactone is an endogenous metabolite and precursor to ascorbic acid, supplied as a 10 mM aqueous solution (1 mL) intended for research and analytical applications. It is provided as an analytical standard for qualitative and quantitative assays and is compatible with HPLC, GC, and MS workflows.
  • Analytical standard quality suitable for HPLC, GC, and MS
  • Provides a stable precursor for ascorbic acid biosynthesis studies
  • Ready-to-use aqueous solution for convenient experimental setup
  • Small vial volume minimizes waste for low-volume assays
  • Intended for research and analytical applications
Catalog No. 50-003-00801 Supplier Medchemexpress LLC Supplier No. HYW01662810MM/1ML
L-Gulono-1,4-lactone is an endogenous metabolite and precursor to ascorbic acid, supplied as a 10 mM aqueous solution (1 mL) intended for research and analytical applications. It is provided as an analytical standard for qualitative and quantitative assays and is compatible with HPLC, GC, and MS workflows.
  • Analytical standard quality suitable for HPLC, GC, and MS
  • Provides a stable precursor for ascorbic acid biosynthesis studies
  • Ready-to-use aqueous solution for convenient experimental setup
  • Small vial volume minimizes waste for low-volume assays
  • Intended for research and analytical applications