Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-gulono-1,4-lactone | 1128-23-0 | MFCD00064331 | 95.0% | 178.14 g·mol⁻¹ | C6H10O6 | 500 MG
SDP

Catalog No. 5000301111
Encompass_Preferred

L-gulono-1,4-lactone is the lactone form of L-gulonic acid and a substrate in the biosynthesis of L-ascorbic acid (vitamin C). Supplied as an analytical standard for research and analytical applications, it is used in metabolic, enzymatic, and analytical studies where a defined endogenous metabolite reference is required.

  • Analytical standard for research and analytical use.
  • Endogenous metabolite and substrate in vitamin C biosynthesis.
  • Molecular formula C6H10O6.
  • Molecular weight 178.14 g·mol⁻¹.
  • CAS number 1128-23-0.
  • Soluble in water at 50 mg/mL; ultrasonic agitation recommended.
  • Available in multiple pack sizes, including 500 mg.

Catalog No. 50-003-01111 Supplier Medchemexpress LLC Supplier No. HYW016628500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-gulono-1,4-lactone is the lactone form of L-gulonic acid and a substrate in the biosynthesis of L-ascorbic acid (vitamin C). Supplied as an analytical standard for research and analytical applications, it is used in metabolic, enzymatic, and analytical studies where a defined endogenous metabolite reference is required.

  • Analytical standard for research and analytical use.
  • Endogenous metabolite and substrate in vitamin C biosynthesis.
  • Molecular formula C6H10O6.
  • Molecular weight 178.14 g·mol⁻¹.
  • CAS number 1128-23-0.
  • Soluble in water at 50 mg/mL; ultrasonic agitation recommended.
  • Available in multiple pack sizes, including 500 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.