Learn More
Medchemexpress LLC HY-W016628 1g Medchemexpress, L-Gulono-1,4-lactone CAS:1128-23-0 Purity:>98%
SDP
Catalog No. 501934619
Encompass_Preferred
Medchemexpress, HY-W016628 1g L-Gulono-1,4-lactone CAS:1128-23-0 L-Gulono-1,4-lactone is a substrate of L-gulono-1,4-lactone oxidoreductase, which catalyzes the last step of the biosynthesis of L-ascorbic (Vatamin) C. In other words, L-Gulono-1,4-lactone is a direct precursor of vitamin C in animals, in plants and in some protists. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4619 Supplier Medchemexpress LLC Supplier No. HYW0166281G
Medchemexpress, HY-W016628 1g L-Gulono-1,4-lactone CAS:1128-23-0 L-Gulono-1,4-lactone is a substrate of L-gulono-1,4-lactone oxidoreductase, which catalyzes the last step of the biosynthesis of L-ascorbic (Vatamin) C. In other words, L-Gulono-1,4-lactone is a direct precursor of vitamin C in animals, in plants and in some protists. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.