Learn More
Selleck Chemical LLC L-Gulono-1 4-lactone
SDP
Catalog No. 5000002944
Encompass_Preferred
L-Gulono-1 4-lactone (L-Gulonolactone L-Gulono-gamma-lactone reduced ascorbic acid) is the substrate of the enzyme L-gulono-1 4-lactone oxidoreductase which catalyzes the last step of the biosynthesis of L-ascorbic acid (vitamin C) in plants and animals
Catalog No. 50-000-02944 Supplier Selleck Chemical LLC Supplier No. S618225MG
L-Gulono-1 4-lactone (L-Gulonolactone L-Gulono-gamma-lactone reduced ascorbic acid) is the substrate of the enzyme L-gulono-1 4-lactone oxidoreductase which catalyzes the last step of the biosynthesis of L-ascorbic acid (vitamin C) in plants and animals