Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Gulose | 6027-89-0 | 95.0% | 10 MG
SDP

Catalog No. 5000338985
Encompass_Preferred

L-Gulose is an L-hexose sugar, the putative furanose form of L-sorbosone, acting as an intermediate in the biosynthesis of L-Ascorbate (vitamin C). This product is intended for research use only and is classified as an endogenous metabolite, listed under saccharides and monosaccharides.

  • 95.0% purity
  • Functions as an intermediate in vitamin C biosynthesis
  • Soluble in H₂O at 250 mg/mL, requiring sonication
  • Soluble in PBS at 100 mg/mL, forming a clear solution and requiring sonication
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months; -20°C for 1 month, protected from light

Catalog No. 50-003-38985 Supplier Medchemexpress LLC Supplier No. HY12839410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Gulose is an L-hexose sugar, the putative furanose form of L-sorbosone, acting as an intermediate in the biosynthesis of L-Ascorbate (vitamin C). This product is intended for research use only and is classified as an endogenous metabolite, listed under saccharides and monosaccharides.

  • 95.0% purity
  • Functions as an intermediate in vitamin C biosynthesis
  • Soluble in H₂O at 250 mg/mL, requiring sonication
  • Soluble in PBS at 100 mg/mL, forming a clear solution and requiring sonication
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months; -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.