Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Gulose | 6027-89-0 | 95.0% | 100 MG
SDP

Catalog No. 5000339975
Encompass_Preferred

L-Gulose is an L-hexose sugar, serving as an intermediate in the biosynthesis of L-Ascorbate (vitamin C). It is also the putative furanose form of L-sorbosone and is classified as an endogenous metabolite, appearing as an off-white to light yellow solid.

  • Classified as a saccharide and monosaccharide.
  • Soluble in H2O: 250 mg/mL (requires ultrasonic).
  • Recommended storage: 4°C, protected from light.
  • For research use only.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protected from light).

Catalog No. 50-003-39975 Supplier Medchemexpress LLC Supplier No. HY128394100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Gulose is an L-hexose sugar, serving as an intermediate in the biosynthesis of L-Ascorbate (vitamin C). It is also the putative furanose form of L-sorbosone and is classified as an endogenous metabolite, appearing as an off-white to light yellow solid.

  • Classified as a saccharide and monosaccharide.
  • Soluble in H2O: 250 mg/mL (requires ultrasonic).
  • Recommended storage: 4°C, protected from light.
  • For research use only.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protected from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.