Learn More
Medchemexpress LLC L-gulose | 6027-89-0 | MFCD00136022 | 95.0% | 180.16 g/mol | C6H12O6 | 5 MG
SDP
Catalog No. 5000264353
Encompass_Preferred
L-Gulose is an L-hexose sugar (the putative furanose form of L-sorbosone) and an intermediate in the biosynthesis of L-ascorbate (vitamin C). Supplied as a research-grade solid, it is intended for metabolic, biochemical, and analytical applications.
  • Rare aldohexose sugar and metabolic intermediate
  • Molecular formula C6H12O6 and molecular weight 180.16 g/mol
  • Typical purity around 95.0%
  • Available in small milligram quantities for analytical use
  • Suitable as a reference standard for pathway and metabolomics studies
  • White to off-white solid form for easy handling and storage
Catalog No. 50-002-64353 Supplier Medchemexpress LLC Supplier No. HY1283945MG
L-Gulose is an L-hexose sugar (the putative furanose form of L-sorbosone) and an intermediate in the biosynthesis of L-ascorbate (vitamin C). Supplied as a research-grade solid, it is intended for metabolic, biochemical, and analytical applications.
  • Rare aldohexose sugar and metabolic intermediate
  • Molecular formula C6H12O6 and molecular weight 180.16 g/mol
  • Typical purity around 95.0%
  • Available in small milligram quantities for analytical use
  • Suitable as a reference standard for pathway and metabolomics studies
  • White to off-white solid form for easy handling and storage