Learn More
Bellco Glass, Inc. L/H Stainless Steel Pitch Blade Assembly , 10L For 10L Bioreator Systems
SDP
Catalog No. 501944734
Encompass_Preferred
SST Pitch Blade Assyembly. MADE IN THE USA 316l stainless steel construction, electropolish finishFor 10L Bioreactor Systems - English ThreadsLeft hand - downOD: 3/1/4"Usually requires 2ea.
Catalog No. 50-194-4734 Supplier Bellco Glass, Inc. Supplier No. 196402220
SST Pitch Blade Assyembly. MADE IN THE USA 316l stainless steel construction, electropolish finishFor 10L Bioreactor Systems - English ThreadsLeft hand - downOD: 3/1/4"Usually requires 2ea.