Learn More
Medchemexpress LLC L-hercynine-d3 | 1639966-63-4 | 99.8% | C9H12D3N3O2 | 10MG
SDP
Catalog No. 5000260335
Encompass_Preferred
L-Hercynine-d3 is a deuterium-labeled stable isotope of L-hercynine supplied as a high-purity solid for use as an internal standard, tracer, or reference in analytical and research applications such as LC-MS, GC-MS, and NMR.
  • Deuterium-labeled analog of L-hercynine.
  • High stated purity of 99.8%.
  • Formulated as a white to light yellow solid suitable for handling and storage.
  • Offered in small milligram pack sizes for analytical use.
  • Documentation available: data sheet, SDS, and COA for quality verification.
Catalog No. 50-002-60335 Supplier Medchemexpress LLC Supplier No. HY143547S10MG
L-Hercynine-d3 is a deuterium-labeled stable isotope of L-hercynine supplied as a high-purity solid for use as an internal standard, tracer, or reference in analytical and research applications such as LC-MS, GC-MS, and NMR.
  • Deuterium-labeled analog of L-hercynine.
  • High stated purity of 99.8%.
  • Formulated as a white to light yellow solid suitable for handling and storage.
  • Offered in small milligram pack sizes for analytical use.
  • Documentation available: data sheet, SDS, and COA for quality verification.