Learn More
Medchemexpress LLC L-Hercynine-d3 | 1639966-63-4 | 99.8% | C9H12D3N3O2 | 2.5 MG
SDP
Catalog No. 5000354926
Encompass_Preferred
L-Hercynine-d3 is the deuterium labeled L-Hercynine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-54926 Supplier Medchemexpress LLC Supplier No. HY143547S2.5MG
L-Hercynine-d3 is the deuterium labeled L-Hercynine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles of drugs