Learn More
Medchemexpress LLC L-Hexanoylcarnitine | 22671-29-0 | 99.9% | 259.34 | 1 ML
SDP
Catalog No. 5000435824
Encompass_Preferred
L-Hexanoylcarnitine is an acylcarnitine associated with celiac disease. It is intended for research use only and is not sold to patients.
  • Acylcarnitine
  • Associated with celiac disease
  • For research use only
  • Classified as a human endogenous metabolite
Catalog No. 50-004-35824 Supplier Medchemexpress LLC Supplier No. HY11314410MM/1ML
L-Hexanoylcarnitine is an acylcarnitine associated with celiac disease. It is intended for research use only and is not sold to patients.
  • Acylcarnitine
  • Associated with celiac disease
  • For research use only
  • Classified as a human endogenous metabolite