Learn More
Medchemexpress LLC L-Hexanoylcarnitine | 22671-29-0 | 99.9% | 259.34 | 100 MG
SDP
Catalog No. 5000436730
Encompass_Preferred
L-Hexanoylcarnitine is an acylcarnitine associated with celiac disease and functions as a human endogenous metabolite. It appears as an off-white to light yellow solid.
  • Associated with celiac disease
  • Functions as a human endogenous metabolite
  • Off-white to light yellow solid appearance
  • For research use only
Catalog No. 50-004-36730 Supplier Medchemexpress LLC Supplier No. HY113144100MG
L-Hexanoylcarnitine is an acylcarnitine associated with celiac disease and functions as a human endogenous metabolite. It appears as an off-white to light yellow solid.
  • Associated with celiac disease
  • Functions as a human endogenous metabolite
  • Off-white to light yellow solid appearance
  • For research use only