Learn More
Medchemexpress LLC L-Hexanoylcarnitine-d3 chloride | 2483831-95-2 | 99.0% | 298.82 | 1 MG
SDP
Catalog No. 5000435671
Encompass_Preferred
L-Hexanoylcarnitine-d3 (chloride) is a deuterium labeled compound designed for quantitative analysis. It serves as a tracer or an internal standard in various analytical techniques.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-35671 Supplier Medchemexpress LLC Supplier No. HY113144AS1MG
L-Hexanoylcarnitine-d3 (chloride) is a deuterium labeled compound designed for quantitative analysis. It serves as a tracer or an internal standard in various analytical techniques.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS