Learn More
Medchemexpress LLC HY-N0832 1g Medchemexpress, L-Hisidine CAS:71-00-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08321G
Encompass_Preferred
Medchemexpress, HY-N0832 1g L-Hisidine CAS:71-00-1 L-Hisidine is an essential amino acid for infants. L-Hisidine is an inhibitor of mitochondrial glutamine transport . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3737