Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Histidine ReagentPlus(R), >=99% (TLC) | 71-00-1 | MFCD00064315 | 500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H8000500G
Encompass
L-Histidine ReagentPlus(R), >=99% (TLC) | Purity: >=99% (TLC) | Mol Wt: 155.15 | 71-00-1 | MFCD00064315 | 500G
Catalog No. NC1264306