Learn More
U.S. Pharmacopeia L-Histidine, 71-00-1, MFCD00064315, 200mg
Supplier:  U.S. Pharmacopeia 1308505
Encompass
Molecular formula C6-H9-N3-O2, Molecular weight 155.16 g/mol, Melting Point 530.6 - 548.6 °F (277 - 287 °C) (decomposes), Synonyms: Glyoxaline-5-alanine
Catalog No. NC9893130