Learn More
Medchemexpress LLC L-Histidine | 71-00-1 | 10MM 1ML
SDP
Catalog No. 5000416927
Encompass_Preferred
L-Histidine is an essential amino acid for infants and an inhibitor of mitochondrial glutamine transport. It is classified as an amino acid and endogenous metabolite. This product is applicable for research in metabolic disease, inflammation/immunology, and other disease research fields. It can also act as a freeze-drying protective agent and a solubilizing agent.
  • Amino acid
  • Endogenous metabolite
  • Metabolic disease research
  • Inflammation/immunology research
  • Disease research fields
  • Freeze-drying protective agent
  • Solubilizing agent
Catalog No. 50-004-16927 Supplier Medchemexpress LLC Supplier No. HYN083210MM/1ML
L-Histidine is an essential amino acid for infants and an inhibitor of mitochondrial glutamine transport. It is classified as an amino acid and endogenous metabolite. This product is applicable for research in metabolic disease, inflammation/immunology, and other disease research fields. It can also act as a freeze-drying protective agent and a solubilizing agent.
  • Amino acid
  • Endogenous metabolite
  • Metabolic disease research
  • Inflammation/immunology research
  • Disease research fields
  • Freeze-drying protective agent
  • Solubilizing agent