Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Histidine benzyl ester bistosylate | 24593-59-7 | 99.86% | 589.68 | 5 G
SDP

Catalog No. 5000349860
Encompass_Preferred

L-Histidine benzyl ester bistosylate is a chemical compound that could play a role in the activation of HutP (an RNA-binding protein). This product is supplied as a white to off-white solid.

  • High purity: 99.86%
  • Molecular weight: 589.68
  • Soluble in DMSO and water
  • Potential role in bacterial infection research

Catalog No. 50-003-49860 Supplier Medchemexpress LLC Supplier No. HY1386525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Histidine benzyl ester bistosylate is a chemical compound that could play a role in the activation of HutP (an RNA-binding protein). This product is supplied as a white to off-white solid.

  • High purity: 99.86%
  • Molecular weight: 589.68
  • Soluble in DMSO and water
  • Potential role in bacterial infection research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.