Learn More
Medchemexpress LLC L-histidine-d5 hydrochloride hydrate | 2483831-75-8 | 99.6% | 214.66 | C6H7D5ClN3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW014423S45MG
Encompass_Preferred
L-histidine-d5 hydrochloride hydrate is a deuterium-labeled amino acid supplied as the hydrochloride hydrate salt for analytical and research use, including mass spectrometry, metabolic tracing, and isotope-labeling studies.
  • Deuterium-labeled L-histidine for isotope tracing and internal standard use.
  • Suitable for mass spectrometry and quantitative assays.
  • High chemical purity (>99.6%).
  • White to off-white solid, easy to handle in the laboratory.
  • Store sealed, away from moisture and light for stability.
Catalog No. 50-003-02728