Learn More
Medchemexpress LLC L-histidine-d5 hydrochloride hydrate | 2483831-75-8 | 99.6% | 214.66 g/mol | C6H7D5ClN3O3 | 1 MG
SDP
Catalog No. 5000303941
Encompass_Preferred
L-histidine-d5 hydrochloride hydrate is a deuterium-labeled L-histidine hydrochloride monohydrate supplied as a research-grade analytical standard. The d5 isotopic label provides a mass shift for use as an internal standard or tracer in mass spectrometry and NMR, and the hydrochloride hydrate form offers good stability and solubility for analytical workflows.
  • Isotopically labeled with five deuterium atoms (d5) for mass-shifted internal standard use.
  • High purity (99.63%) suitable for quantitative and qualitative analyses.
  • Hydrochloride hydrate form enhances stability and solubility.
  • Supplied in milligram-scale quantities for method development and calibration.
  • Appropriate for mass spectrometry, NMR, and tracer studies in research applications.
Catalog No. 50-003-03941 Supplier Medchemexpress LLC Supplier No. HYW014423S41MG
L-histidine-d5 hydrochloride hydrate is a deuterium-labeled L-histidine hydrochloride monohydrate supplied as a research-grade analytical standard. The d5 isotopic label provides a mass shift for use as an internal standard or tracer in mass spectrometry and NMR, and the hydrochloride hydrate form offers good stability and solubility for analytical workflows.
  • Isotopically labeled with five deuterium atoms (d5) for mass-shifted internal standard use.
  • High purity (99.63%) suitable for quantitative and qualitative analyses.
  • Hydrochloride hydrate form enhances stability and solubility.
  • Supplied in milligram-scale quantities for method development and calibration.
  • Appropriate for mass spectrometry, NMR, and tracer studies in research applications.