Learn More
Medchemexpress LLC L-histidine-d5 hydrochloride hydrate | 2483831-75-8 | 99.6% | 214.66 g·mol⁻¹ | C6H7D5ClN3O3 | 10 MG
SDP
Catalog No. 5000303943
Encompass_Preferred
L-histidine-d5 hydrochloride hydrate is the deuterium-labeled form of the amino acid L-histidine supplied as the hydrochloride hydrate salt. It is intended as a high-purity analytical standard for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS workflows.
  • Deuterium-labeled (d5) for isotopic tracing.
  • High purity suitable for analytical applications.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in small pack sizes for method development.
  • Useful as a tracer in metabolic and quantitative studies.
Catalog No. 50-003-03943 Supplier Medchemexpress LLC Supplier No. HYW014423S410MG
L-histidine-d5 hydrochloride hydrate is the deuterium-labeled form of the amino acid L-histidine supplied as the hydrochloride hydrate salt. It is intended as a high-purity analytical standard for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS workflows.
  • Deuterium-labeled (d5) for isotopic tracing.
  • High purity suitable for analytical applications.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in small pack sizes for method development.
  • Useful as a tracer in metabolic and quantitative studies.