Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-histidine-d5 hydrochloride hydrate | 2483831-75-8 | 99.6% | 214.66 g/mol | C6H7D5ClN3O3 | 100 MG
SDP

Catalog No. 5000303305
Encompass_Preferred

L-histidine-d5 hydrochloride hydrate is the deuterium-labeled form of L-histidine hydrochloride hydrate supplied as a hydrochloride hydrate salt. It is intended for analytical and research use as an isotopically labeled standard or tracer for mass spectrometry and metabolic studies, enabling accurate quantitation and isotope-tracing experiments.

  • Deuterium-labeled amino acid useful for isotope tracing and metabolic studies.
  • High purity (99.63%) suitable for analytical applications.
  • Molecular formula C6H7D5ClN3O3 and molecular weight 214.66 g/mol.
  • Available in small pack sizes appropriate for standards (1 mg-500 mg).
  • Supplied as a hydrochloride hydrate for enhanced solubility and stability.
  • Suitable as an internal standard for quantitative mass spectrometry.

Catalog No. 50-003-03305 Supplier Medchemexpress LLC Supplier No. HYW014423S4100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-histidine-d5 hydrochloride hydrate is the deuterium-labeled form of L-histidine hydrochloride hydrate supplied as a hydrochloride hydrate salt. It is intended for analytical and research use as an isotopically labeled standard or tracer for mass spectrometry and metabolic studies, enabling accurate quantitation and isotope-tracing experiments.

  • Deuterium-labeled amino acid useful for isotope tracing and metabolic studies.
  • High purity (99.63%) suitable for analytical applications.
  • Molecular formula C6H7D5ClN3O3 and molecular weight 214.66 g/mol.
  • Available in small pack sizes appropriate for standards (1 mg-500 mg).
  • Supplied as a hydrochloride hydrate for enhanced solubility and stability.
  • Suitable as an internal standard for quantitative mass spectrometry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.